The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029498.6 | NP_001024669.1 | 3 - 4 | 1158 | 83 |
NM_001373594.3 | NP_001359567.1 | 4 - 4 | 1208 | 83 | |
NM_001373595.3 | NP_001360748.1 | 3 - 4 | 635 | 83 | |
NM_001375178.1 | NP_001362050.1 | 2 - 3 | 565 | 83 | |
NM_001381166.1 | NP_001367127.1 | 3 - 4 | 1158 | 83 | |
NM_001383681.1 | 2 - 3 | 565 | 83 | ||
NM_182447.10 | NP_872247.4 | 2 - 3 | 570 | 83 | |
GenBank mRNA | AY836556.1 | - | 3 - 4 | 1123 | 83 |
AY836557.1 | - | 4 - 5 | 665 | 83 | |
AY836558.1 | - | 3 - 4 | 1208 | 83 |