Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ser-2
Entrez Gene ID
182110
Gene Name
protein-coding
Gene Aliases
CELE_C02D4.2
Chromosome Location
Chr.X: 15286533 - 15302963 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029164.3 NP_001024335.1 10 - 11 1146 130
NM_001029165.5 NP_001024336.1 10 - 11 1622 130
NM_001029168.4 NP_001024339.1 10 - 11 1215 130
NM_001029169.5 NP_001024340.1 9 - 10 1231 130
NM_001270331.4 NP_001257260.1 9 - 10 1110 130
GenBank mRNA GQ217520.1 - 9 - 10 1113 130

View Full Product Details