Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
svh-5
Entrez Gene ID
181627
Gene Name
protein-coding
Gene Aliases
CELE_C33A11.4
Chromosome Location
Chr.X: 15369974 - 15377824 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270332.3 NP_001257261.1 8 - 9 1503 124
NM_001383683.1 9 - 10 1750 124

View Full Product Details