Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
psa-3
Entrez Gene ID
181631
Gene Name
protein-coding
Gene Aliases
CELE_F39D8.2
Chromosome Location
Chr.X: 15407931 - 15419298 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083296.5 NP_001076765.2 2 - 3 288 58
NM_001083297.5 NP_001076766.1 2 - 3 294 58
NM_001270334.3 NP_001257263.1 1 - 2 90 58
GenBank mRNA DQ118141.1 - 2 - 3 186 58

View Full Product Details