Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-36
Entrez Gene ID
181630
Gene Name
protein-coding
Gene Aliases
CELE_F39D8.1
Chromosome Location
Chr.X: 15409148 - 15415493 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001364051.5 NP_001350994.1 9 - 9 2046 113
NM_001364052.5 NP_001350995.1 9 - 9 2046 113
NM_001364053.4 NP_001350996.1 9 - 10 2061 113
NM_001364054.4 NP_001350997.1 9 - 10 2061 113
NM_078144.4 NP_510545.1 7 - 8 2849 113
NM_078145.5 NP_510546.1 7 - 8 2882 113
NM_078146.5 NP_510547.1 7 - 8 2592 113

View Full Product Details