Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nas-13
Entrez Gene ID
185492
Gene Name
Zinc metalloproteinase nas-13
Gene Aliases
CELE_F39D8.4
Chromosome Location
Chr.X: 15425099 - 15428253 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078148.6 NP_510549.3 4 - 5 969 85

View Full Product Details