Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nkat-3
Entrez Gene ID
181633
Gene Name
protein-coding
Gene Aliases
CELE_R03A10.4
Chromosome Location
Chr.X: 15441729 - 15443630 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029651.5 NP_001024822.1 4 - 5 1133 71
NM_001029652.6 NP_001024823.1 4 - 5 1061 71

View Full Product Details