Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gob-1
Entrez Gene ID
181637
Gene Name
Trehalose-phosphatase
Gene Aliases
CELE_H13N06.3
Chromosome Location
Chr.X: 15493602 - 15497504 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078159.5 NP_510560.2 2 - 3 146 103

View Full Product Details