Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eat-20
Entrez Gene ID
181641
Gene Name
protein-coding
Gene Aliases
CELE_H30A04.1
Chromosome Location
Chr.X: 15510011 - 15524030 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029570.6 NP_001024741.1 10 - 11 1383 76
NM_001383685.1 11 - 12 1435 76
NR_002425.1 10 - 11 1285 76
GenBank mRNA AB032748.1 - 10 - 11 1381 76
AB032749.1 - 10 - 11 1381 76

View Full Product Details