Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
olrn-1
Entrez Gene ID
181642
Gene Name
protein-coding
Gene Aliases
CELE_C02C6.2
Chromosome Location
Chr.X: 15537662 - 15546712 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029163.7 NP_001024334.1 3 - 4 1207 67
NM_001373597.2 NP_001359691.1 3 - 4 1193 67

View Full Product Details