Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dyn-1
Entrez Gene ID
181644
Gene Name
protein-coding
Gene Aliases
CELE_C02C6.1
Chromosome Location
Chr.X: 15568884 - 15573699 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373600.3 NP_001359587.1 7 - 8 2478 79

View Full Product Details