Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
erv-46
Entrez Gene ID
181652
Gene Name
yeast ERV (ER to Golgi transport Vesicle protein) homolog
Gene Aliases
CELE_K09E9.2
Chromosome Location
Chr.X: 15613797 - 15616519 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383686.1 4 - 5 552 114

View Full Product Details