Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-15A
Entrez Gene ID
181663
Gene Name
Protein lin-15A
Gene Aliases
CELE_ZK678.1
Chromosome Location
Chr.X: 15731968 - 15735371 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078187.7 NP_510588.1 1 - 2 288 99
GenBank mRNA U10411.1 - 1 - 2 310 99

View Full Product Details