Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
wrt-4
Entrez Gene ID
181664
Gene Name
Warthog protein 4 N-product
Gene Aliases
CELE_ZK678.5
Chromosome Location
Chr.X: 15752253 - 15757269 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078192.8 NP_510593.1 1 - 2 106 73

View Full Product Details