Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acl-1
Entrez Gene ID
181671
Gene Name
Putative 1-acyl-sn-glycerol-3-phosphate acyltransferase acl-1
Gene Aliases
CELE_F59F4.4
Chromosome Location
Chr.X: 15827018 - 15852001 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078205.8 NP_510606.1 5 - 6 604 57

View Full Product Details