Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ifa-3
Entrez Gene ID
181699
Gene Name
Intermediate filament protein ifa-3
Gene Aliases
CELE_F52E10.5
Chromosome Location
Chr.X: 16267197 - 16270069 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078248.10 NP_510649.3 1 - 2 500 119

View Full Product Details