Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
twk-21
Entrez Gene ID
192078
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_T01B4.1
Chromosome Location
Chr.X: 16299892 - 16304191 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078253.3 NP_510654.2 6 - 7 972 69

View Full Product Details