Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-22
Entrez Gene ID
181703
Gene Name
protein-coding
Gene Aliases
CELE_T01B4.2
Chromosome Location
Chr.X: 16306464 - 16314998 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001375199.1 NP_001361830.1 1 - 2 186 106

View Full Product Details