Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- nas-39
- Entrez Gene ID
-
3565986
- Gene Name
- Zinc metalloproteinase nas-39
- Gene Aliases
-
CELE_F38E9.2
- Chromosome Location
-
Chr.X: 16456927 - 16474601 on Build WBcel235
- Gene Symbol
- F38E9.1
- Entrez Gene ID
-
181712
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_F38E9.1
- Chromosome Location
-
Chr.X: 16464538 - 16474994 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078272.7 |
NP_510673.1 |
5 - 6 |
818 |
67 |
View Full Product Details