Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nas-39
Entrez Gene ID
3565986
Gene Name
Zinc metalloproteinase nas-39
Gene Aliases
CELE_F38E9.2
Chromosome Location
Chr.X: 16456927 - 16474601 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373610.3 NP_001360030.1 3 - 4 468 76

View Full Product Details