Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cah-3
Entrez Gene ID
181713
Gene Name
Putative carbonic anhydrase 3
Gene Aliases
CELE_K05G3.3
Chromosome Location
Chr.X: 16478452 - 16489396 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383694.1 4 - 5 518 133

View Full Product Details