Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hda-4
Entrez Gene ID
181723
Gene Name
protein-coding
Gene Aliases
CELE_C10E2.3
Chromosome Location
Chr.X: 16729559 - 16744769 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270349.3 NP_001257278.1 9 - 10 2386 61
NM_001270350.5 NP_001257279.1 9 - 10 2388 61

View Full Product Details