Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aak-2
Entrez Gene ID
181727
Gene Name
protein-coding
Gene Aliases
CELE_T01C8.1
Chromosome Location
Chr.X: 16796087 - 16803404 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029697.5 NP_001024868.1 5 - 6 959 76
NM_078310.7 NP_510711.2 6 - 7 1104 76
GenBank mRNA AY347273.1 - 6 - 7 1102 76

View Full Product Details