Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mec-4
Entrez Gene ID
181728
Gene Name
Degenerin mec-4
Gene Aliases
CELE_T01C8.7
Chromosome Location
Chr.X: 16803321 - 16807115 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078311.8 NP_510712.2 10 - 11 1735 105

View Full Product Details