Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tps-1
Entrez Gene ID
181778
Gene Name
protein-coding
Gene Aliases
CELE_ZK54.2
Chromosome Location
Chr.X: 17467121 - 17474585 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047837.4 NP_001041302.1 7 - 8 1202 71
NM_001047838.4 NP_001041303.1 6 - 7 1034 71
GenBank mRNA AJ512332.1 - 7 - 8 1145 71
AJ811573.1 - 7 - 8 1173 71

View Full Product Details