Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nhr-1
Entrez Gene ID
181783
Gene Name
protein-coding
Gene Aliases
CELE_R09G11.2
Chromosome Location
Chr.X: 17514063 - 17528197 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029682.5 NP_001024853.1 3 - 4 383 76
NM_001029684.6 NP_001024855.1 2 - 3 280 76
NM_001381184.1 NP_001367802.1 1 - 2 188 76
GenBank mRNA AY204163.1 - 2 - 3 264 76
AY204164.1 - 2 - 3 278 76
AY204165.1 - 2 - 3 278 76

View Full Product Details