Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aps-3
Entrez Gene ID
171691
Gene Name
Clat_adaptor_s domain-containing protein
Gene Aliases
CELE_Y48G8AL.14
Chromosome Location
Chr.I: 1154568 - 1156020 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381075.1 NP_001368004.1 1 - 2 273 148

View Full Product Details