Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lin-65
Entrez Gene ID
171739
Gene Name
protein-coding
Gene Aliases
CELE_Y71G12B.9
Chromosome Location
Chr.I: 1755329 - 1766400 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170802.6 NP_740785.2 3 - 4 1058 152
NM_170803.6 NP_740786.3 3 - 4 1057 152
GenBank mRNA AY818712.1 - 3 - 4 1080 152

View Full Product Details