Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tpxl-1
Entrez Gene ID
171786
Gene Name
protein-coding
Gene Aliases
CELE_Y39G10AR.12
Chromosome Location
Chr.I: 2297280 - 2304397 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170821.6 NP_740805.1 2 - 3 1188 55
NM_170822.8 NP_740806.1 2 - 3 1192 55
GenBank mRNA DQ178245.1 - 2 - 3 1188 55
DQ178246.1 - 2 - 3 1188 55

View Full Product Details