Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lpd-3
Entrez Gene ID
171926
Gene Name
protein-coding
Gene Aliases
CELE_Y47G6A.23
Chromosome Location
Chr.I: 3564963 - 3600735 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313535.2 NP_001300464.1 11 - 12 2519 112
NM_001313536.3 NP_001300465.1 11 - 12 2529 112
NM_001313537.3 NP_001300466.1 11 - 12 2465 112
NM_001313538.2 NP_001300467.1 11 - 12 2523 112

View Full Product Details