The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313535.2 | NP_001300464.1 | 5 - 6 | 921 | 72 |
NM_001313536.3 | NP_001300465.1 | 5 - 6 | 931 | 72 | |
NM_001313537.3 | NP_001300466.1 | 5 - 6 | 867 | 72 | |
NM_001313538.2 | NP_001300467.1 | 5 - 6 | 925 | 72 |