Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lpd-3
Entrez Gene ID
171926
Gene Name
protein-coding
Gene Aliases
CELE_Y47G6A.23
Chromosome Location
Chr.I: 3564963 - 3600735 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313535.2 NP_001300464.1 8 - 9 1776 104
NM_001313536.3 NP_001300465.1 8 - 9 1786 104
NM_001313537.3 NP_001300466.1 8 - 9 1722 104
NM_001313538.2 NP_001300467.1 8 - 9 1780 104

View Full Product Details