The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001374889.1 | NP_001361919.1 | 14 - 15 | 2096 | 140 |
NM_001374890.2 | NP_001362173.1 | 14 - 15 | 2100 | 140 |