Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eat-16
Entrez Gene ID
172823
Gene Name
protein-coding
Gene Aliases
CELE_C16C2.2
Chromosome Location
Chr.I: 9722716 - 9726288 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170911.5 NP_740904.1 6 - 7 779 71
NM_170912.6 NP_740905.1 6 - 7 784 71

View Full Product Details