Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scpl-1
Entrez Gene ID
181944
Gene Name
protein-coding
Gene Aliases
CELE_B0379.4
Chromosome Location
Chr.I: 10085703 - 10102012 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128939.2 NP_001122411.1 3 - 4 419 103
NM_001383711.1 7 - 8 780 103
NM_170918.6 NP_740912.2 6 - 7 1191 103

View Full Product Details