Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bbs-1
Entrez Gene ID
260219
Gene Name
Bardet-Biedl syndrome 1 protein homolog
Gene Aliases
CELE_Y105E8A.5
Chromosome Location
Chr.I: 14371138 - 14376086 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170937.3 NP_740933.1 8 - 9 1546 84

View Full Product Details