Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ipla-1
Entrez Gene ID
173795
Gene Name
protein-coding
Gene Aliases
CELE_M03A1.6
Chromosome Location
Chr.II: 4590939 - 4601360 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170979.6 NP_740976.1 1 - 2 348 82

View Full Product Details