Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- shc-2
- Entrez Gene ID
-
173860
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_T27F7.2
- Chromosome Location
-
Chr.II: 4969163 - 4988940 on Build WBcel235
- Gene Symbol
- T27F7.12
- Entrez Gene ID
-
13184806
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_T27F7.12
- Chromosome Location
-
Chr.II: 4987105 - 4990836 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383825.1 |
|
2 - 3 |
432 |
122 |
View Full Product Details