Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
osta-2
Entrez Gene ID
173966
Gene Name
Organic solute transporter alpha-like protein 2
Gene Aliases
CELE_C18A3.4
Chromosome Location
Chr.II: 5707691 - 5712351 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171004.7 NP_741003.1 3 - 4 432 125
NR_136362.1 3 - 4 432 125
NR_136363.1 3 - 4 432 125

View Full Product Details