Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cgt-3
Entrez Gene ID
174001
Gene Name
protein-coding
Gene Aliases
CELE_F59G1.1
Chromosome Location
Chr.II: 5911324 - 5916910 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062780.6 NP_495181.2 2 - 3 176 58
NR_151564.1 1 - 2 69 58
GenBank mRNA DQ178633.1 - - 176 58
DQ178634.1 - - 116 58

View Full Product Details