Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-44
Entrez Gene ID
174005
Gene Name
protein-coding
Gene Aliases
CELE_F28B12.2
Chromosome Location
Chr.II: 5928802 - 5940830 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356848.3 NP_001343765.1 5 - 5 535 87
NM_001356850.3 NP_001343767.1 5 - 5 538 87
NM_001356852.3 NP_001343768.1 4 - 5 483 87
NM_001381441.2 NP_001367307.1 4 - 5 503 87
NM_171007.5 NP_741006.3 5 - 6 563 87

View Full Product Details