Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-27
Entrez Gene ID
174121
Gene Name
protein-coding
Gene Aliases
CELE_C04A2.3
Chromosome Location
Chr.II: 6835008 - 6847908 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276731.3 NP_001263660.1 2 - 3 383 73
NM_001313565.3 NP_001300494.1 8 - 8 1106 73
NM_001313566.3 NP_001300495.1 8 - 8 1097 73
NM_001313568.3 NP_001300497.1 7 - 8 1004 73
NM_001313569.3 NP_001300498.1 7 - 8 1004 73
NM_001313570.3 NP_001300499.1 7 - 8 995 73
NM_001313571.3 NP_001300500.1 7 - 8 995 73
NM_001313575.3 NP_001300504.1 5 - 6 674 73
NM_001381461.1 NP_001368508.1 8 - 9 1097 73
NM_001381462.1 NP_001368509.1 5 - 6 683 73
NM_001381463.1 NP_001367420.1 8 - 9 1170 73
NM_001381464.1 NP_001368510.1 2 - 3 392 73
NM_001381465.1 NP_001368511.1 2 - 3 383 73
NM_001383871.1 8 - 9 1150 73
NM_001383872.1 7 - 8 1127 73
NM_001383873.1 8 - 9 1172 73
NM_171011.4 NP_741012.1 8 - 9 1166 73

View Full Product Details