Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnj-5
Entrez Gene ID
174122
Gene Name
protein-coding
Gene Aliases
CELE_C04A2.7
Chromosome Location
Chr.II: 6853458 - 6858301 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171015.6 NP_741016.1 13 - 14 2532 72

View Full Product Details