Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rsp-7
Entrez Gene ID
174646
Gene Name
protein-coding
Gene Aliases
CELE_D2089.1
Chromosome Location
Chr.II: 10659202 - 10662317 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383939.1 2 - 3 280 79
NM_001383940.1 2 - 3 274 79

View Full Product Details