Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-53
Entrez Gene ID
174693
Gene Name
protein-coding
Gene Aliases
CELE_F45E10.1
Chromosome Location
Chr.II: 11058047 - 11089456 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026998.5 NP_001022169.1 15 - 16 2057 85
NM_001026999.8 NP_001022170.1 15 - 16 2057 85
NM_001027000.6 NP_001022171.1 12 - 13 1852 85
NM_001027001.5 NP_001022172.1 7 - 8 797 85
NM_001267342.3 NP_001254271.1 15 - 16 2057 85
NM_001267343.3 NP_001254272.1 15 - 16 2057 85
NM_001267344.3 NP_001254273.1 8 - 9 1133 85
NM_171039.3 NP_741045.1 15 - 16 2098 85

View Full Product Details