Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-35
Entrez Gene ID
189960
Gene Name
protein-coding
Gene Aliases
CELE_Y46G5A.26
Chromosome Location
Chr.II: 12836480 - 12840157 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027267.5 NP_001022438.1 10 - 11 1375 78
NM_001373808.1 NP_001359643.1 11 - 12 1548 78

View Full Product Details