Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gst-34
Entrez Gene ID
260015
Gene Name
Glutathione S-Transferase
Gene Aliases
CELE_Y1H11.1
Chromosome Location
Chr.II: 15179906 - 15183239 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171053.3 NP_741060.2 1 - 2 158 60

View Full Product Details