Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kbp-2
Entrez Gene ID
266885
Gene Name
KNL (kinetochore null) Binding Protein
Gene Aliases
CELE_F26F4.13
Chromosome Location
Chr.III: 4903182 - 4904188 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001346612.3 NP_001333543.1 3 - 3 338 65

View Full Product Details