Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mdt-15
Entrez Gene ID
175817
Gene Name
protein-coding
Gene Aliases
CELE_R12B2.5
Chromosome Location
Chr.III: 5828557 - 5833746 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171134.8 NP_741151.1 3 - 4 546 86

View Full Product Details