Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zmp-1
Entrez Gene ID
175839
Gene Name
Matrix metalloproteinase-A
Gene Aliases
CELE_EGAP1.3
Chromosome Location
Chr.III: 5926895 - 5932953 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171138.3 NP_741156.1 5 - 6 659 110
GenBank mRNA AB007817.1 - 5 - 6 659 110

View Full Product Details